Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CG11147
Entrez Gene ID
33803
Gene Name
CG11147-PA
Gene Aliases
Dmel\CG11147, Dmel_CG11147
Chromosome Location
Chr.2L: 5733675 - 5741244 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031734
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135110.1 NP_608954.1 7 - 8 1824 65
NM_164658.2 NP_723114.1 6 - 7 1697 65
GenBank mRNA BT010210.1 - 7 - 8 1825 65

View Full Product Details