Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
flr
Entrez Gene ID
39505
Gene Name
flare
Gene Aliases
AIP1, Aip1, CG10724, Dmel\CG10724, Dmel_CG10724, Flare, aip1
Chromosome Location
Chr.3L: 13412843 - 13416453 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260049
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140385.4 NP_648642.1 2 - 3 250 69
NM_168543.2 NP_729891.1 2 - 3 185 69
GenBank mRNA AY061340.1 - 2 - 3 231 69
BT099796.1 - 2 - 3 185 69

View Full Product Details