Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abp1
Entrez Gene ID
39520
Gene Name
Actin binding protein 1
Gene Aliases
CG10083, Dmel\CG10083, Dmel_CG10083, abp1
Chromosome Location
Chr.3L: 13483533 - 13486280 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036372
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140400.2 NP_648657.1 2 - 3 1130 58
GenBank mRNA BT021389.1 - 2 - 3 1132 58

View Full Product Details