Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RhoGAP71E
Entrez Gene ID
39693
Gene Name
Rho GTPase activating protein at 71E
Gene Aliases
BcDNA:LD07615, C- GAP, CG16980, CG32149, CG7396, Dmel\CG32149, Dmel_CG32149, RhoGAP-71E1, RhoGap71E, l(3)j6B9, rhogap71e
Chromosome Location
Chr.3L: 15588860 - 15609125 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036518
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144484.3 NP_001137956.1 1 - 2 1139 95
NM_001202183.2 NP_001189112.1 1 - 2 1139 95
NM_001274957.1 NP_001261886.1 1 - 2 1139 95
NM_001274958.1 NP_001261887.1 1 - 2 1139 95
NM_140527.5 NP_648784.1 1 - 2 1139 95
NM_168629.4 NP_730060.1 1 - 2 1139 95
NM_168630.4 NP_730061.1 1 - 2 1139 95
GenBank mRNA AY061070.1 - 1 - 2 1119 95

View Full Product Details