Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGAP71E
Entrez Gene ID
39693
Gene Name
Rho GTPase activating protein at 71E
Gene Aliases
BcDNA:LD07615, C- GAP, CG16980, CG32149, CG7396, Dmel\CG32149, Dmel_CG32149, RhoGAP-71E1, RhoGap71E, l(3)j6B9, rhogap71e
Chromosome Location
Chr.3L: 15588860 - 15609125 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036518
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144484.3 NP_001137956.1 4 - 5 1591 115
NM_001202183.2 NP_001189112.1 4 - 5 1591 115
NM_140527.5 NP_648784.1 4 - 5 1591 115
NM_168629.4 NP_730060.1 4 - 5 1591 115
NM_168630.4 NP_730061.1 4 - 5 1591 115
GenBank mRNA AY061070.1 - 4 - 5 1571 115

View Full Product Details