The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_140636.3 | NP_648893.1 | 5 - 6 | 1216 | 72 |
GenBank mRNA | AY058257.1 | - | 5 - 6 | 1183 | 72 |