Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TyrRS
- Entrez Gene ID
-
39829
- Gene Name
- Tyrosyl-tRNA synthetase
- Gene Aliases
-
Aats-tyr, CG4561, Dmel\CG4561, Dmel_CG4561, EMAP, YARS, YRS, anon-EST:Posey261, dTyrRS, dYARS
- Chromosome Location
-
Chr.3L: 16417349 - 16419302 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027080
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140638.3 |
NP_648895.1 |
3 - 4 |
1394 |
69 |
GenBank mRNA |
AY051662.1 |
- |
3 - 4 |
1387 |
69 |
View Full Product Details