Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lasp
- Entrez Gene ID
-
39864
- Gene Name
- CG3849-PA
- Gene Aliases
-
CG3849, Dmel\CG3849, Dmel_CG3849, anon-73Bd, cDNA B, lasp
- Chromosome Location
-
Chr.3L: 16676406 - 16711677 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0063485
- Gene Symbol
- CG43954
- Entrez Gene ID
-
14462875
- Gene Name
- CG43954-PA
- Gene Aliases
-
Dmel\CG43954, Dmel_CG43954
- Chromosome Location
-
Chr.3L: 16676406 - 16711677 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264605
View Full Product Details