Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGRP-SB2
Entrez Gene ID
39869
Gene Name
Peptidoglycan recognition protein SB2
Gene Aliases
CG9697, Dmel\CG9697, Dmel_CG9697, PGRP-SB
Chromosome Location
Chr.3L: 16726541 - 16727288 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043577
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140659.3 NP_648916.1 1 - 2 370 65

View Full Product Details