Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oatp74D
Entrez Gene ID
39954
Gene Name
Organic anion transporting polypeptide 74D
Gene Aliases
CG7571, Dmel\CG7571, Dmel_CG7571, EcI, oatp 74D
Chromosome Location
Chr.3L: 17487118 - 17511693 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036732
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169989.2 NP_001163460.1 7 - 8 2762 64
NM_001300166.1 NP_001287095.1 7 - 8 2705 64
NM_001300167.1 NP_001287096.1 6 - 7 3110 64
NM_140732.3 NP_648989.1 7 - 8 2751 64
GenBank mRNA BT001436.1 - 7 - 8 2730 64

View Full Product Details