Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps60
Entrez Gene ID
39964
Gene Name
Vacuolar protein sorting 60
Gene Aliases
CG6259, Chmp5, Dchmp5, Dmel\CG6259, Dmel_CG6259, VPS60, anon-74EFb, bs36h07.y1
Chromosome Location
Chr.3L: 17620123 - 17621625 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036740
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275065.1 NP_001261994.1 4 - 5 868 62
NM_140740.4 NP_648997.1 4 - 5 748 62
GenBank mRNA AY071206.1 - 4 - 5 741 62

View Full Product Details