Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir75a
- Entrez Gene ID
-
39982
- Gene Name
- Ionotropic receptor 75a
- Gene Aliases
-
CG14585, DmIr75a, DmelIR75a, Dmel\CG14585, Dmel_CG14585, IR75a, IR75abc, Ir75abc
- Chromosome Location
-
Chr.3L: 17815570 - 17817921 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036757
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140755.3 |
NP_649012.2 |
3 - 4 |
622 |
86 |
GenBank mRNA |
KX694388.1 |
- |
3 - 4 |
622 |
86 |
View Full Product Details