Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir75c
- Entrez Gene ID
-
39983
- Gene Name
- Ionotropic receptor 75c
- Gene Aliases
-
CG14586, CG14586-A, CG42642, DmelIR75c, Dmel\CG42642, Dmel_CG42642, IR75abc, IR75c, Ir75abc
- Chromosome Location
-
Chr.3L: 17820493 - 17822834 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261401
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140756.3 |
NP_649013.3 |
5 - 6 |
1356 |
85 |
GenBank mRNA |
KY205712.1 |
- |
5 - 6 |
1356 |
85 |
View Full Product Details