Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wnd
Entrez Gene ID
40143
Gene Name
wallenda
Gene Aliases
CG8789, DLK, DLK/ZPK, Dmel\CG8789, Dmel_CG8789, MAPKKK, Wnd, Wnd/DLK
Chromosome Location
Chr.3L: 19624406 - 19635783 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036896
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202203.1 NP_001189132.1 6 - 7 3113 71
NM_140880.3 NP_649137.3 7 - 8 2840 71
NM_176362.2 NP_788540.1 7 - 8 2763 71
NM_176363.3 NP_788541.1 7 - 8 2968 71
GenBank mRNA AY094787.1 - 7 - 8 2790 71
BT100072.1 - 6 - 7 3112 71

View Full Product Details