Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Trpml
- Entrez Gene ID
-
40152
- Gene Name
- Transient receptor potential cation channel, mucolipin
- Gene Aliases
-
CG8743, CT25240, DmTRPML, Dmel\CG8743, Dmel_CG8743, TRPML, l(3)76BDi, trpml
- Chromosome Location
-
Chr.3L: 19714418 - 19718328 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0262516
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140888.4 |
NP_649145.1 |
1 - 2 |
623 |
73 |
GenBank mRNA |
AY070709.1 |
- |
1 - 2 |
598 |
73 |
- Gene Symbol
- CG42638
- Entrez Gene ID
-
8673965
- Gene Name
- CG42638-PA
- Gene Aliases
-
Dmel\CG42638, Dmel_CG42638, noncoding_13567
- Chromosome Location
-
Chr.3L: 19745834 - 19749744 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261361
View Full Product Details