Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- gogo
- Entrez Gene ID
-
40225
- Gene Name
- golden goal
- Gene Aliases
-
CG13815, CG13817, CG32227, CT33316, Dmel\CG32227, Dmel_CG32227, Gogo
- Chromosome Location
-
Chr.3L: 20268859 - 20295133 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052227
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140943.3 |
NP_649200.2 |
4 - 5 |
2184 |
83 |
GenBank mRNA |
BT009982.1 |
- |
4 - 5 |
2182 |
83 |
View Full Product Details