Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gogo
Entrez Gene ID
40225
Gene Name
golden goal
Gene Aliases
CG13815, CG13817, CG32227, CT33316, Dmel\CG32227, Dmel_CG32227, Gogo
Chromosome Location
Chr.3L: 20268859 - 20295133 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052227
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140943.3 NP_649200.2 9 - 10 3593 58
GenBank mRNA BT009982.1 - 9 - 10 3591 58

View Full Product Details