Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRG1
Entrez Gene ID
40228
Gene Name
FSHD region gene 1
Gene Aliases
CG6480, DmFRG1, Dmel\CG6480, Dmel_CG6480, dFRG1
Chromosome Location
Chr.3L: 20295802 - 20296971 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036964
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140945.3 NP_649202.1 3 - 4 508 69
GenBank mRNA BT023262.2 - 3 - 4 471 69

View Full Product Details