Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bru1
Entrez Gene ID
34648
Gene Name
bruno 1
Gene Aliases
ARET, Aret, BcDNA:GM15173, Bru, Bruno, CG31762, CG6319, Dm Bru, Dmel\CG31762, Dmel_CG31762, NEST:bs01g04, NEST:bs34h11, aret, aret/bruno1, arrest/Bruno, bru, bru-1, bruno, ms(2)01284
Chromosome Location
Chr.2L: 12175004 - 12313438 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000114
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273476.1 NP_001260405.1 10 - 11 2110 60
NM_165002.2 NP_723737.1 11 - 12 3170 60
NM_165003.3 NP_723738.2 12 - 13 2433 60
NM_165004.3 NP_723739.1 10 - 11 2385 60
GenBank mRNA BT133212.1 - 11 - 12 3149 60

View Full Product Details