Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bru2
Entrez Gene ID
250811
Gene Name
bruno 2
Gene Aliases
Bru-2, CG17008, CG31761, CG31763, CG43065, CG6319, Dm Bru-2, Dmel\CG43065, Dmel_CG43065, bru-2
Chromosome Location
Chr.2L: 12344219 - 12397528 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262475
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042891.3 NP_001036356.1 2 - 3 1413 85
NM_001259069.2 NP_001245998.1 2 - 3 1320 85
NM_001259071.2 NP_001246000.1 2 - 3 1413 85
NM_001273481.1 NP_001260410.1 2 - 3 1696 85
NM_001273482.1 NP_001260411.1 2 - 3 1320 85
NM_135715.4 NP_609559.1 2 - 3 1320 85
NM_165007.3 NP_723742.2 2 - 3 1320 85
NM_176025.3 NP_788039.1 2 - 3 1413 85
GenBank mRNA AY069464.1 - 2 - 3 1287 85

View Full Product Details