Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- bru2
- Entrez Gene ID
-
250811
- Gene Name
- bruno 2
- Gene Aliases
-
Bru-2, CG17008, CG31761, CG31763, CG43065, CG6319, Dm Bru-2, Dmel\CG43065, Dmel_CG43065, bru-2
- Chromosome Location
-
Chr.2L: 12344219 - 12397528 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0262475
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135715.4 |
NP_609559.1 |
7 - 8 |
2335 |
86 |
GenBank mRNA |
AY069464.1 |
- |
7 - 8 |
2302 |
86 |
View Full Product Details