Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vir-1
Entrez Gene ID
34652
Gene Name
virus-induced RNA 1
Gene Aliases
BEST:GH02216, CG15493, CG31764, CG5453, Dmel\CG31764, Dmel_CG31764, Vir-1, Vir1, vir1
Chromosome Location
Chr.2L: 12403270 - 12423439 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043841
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165010.3 NP_723746.1 4 - 5 858 74
NM_165011.3 NP_723747.1 5 - 6 942 74
GenBank mRNA BT044312.1 - 5 - 6 938 74

View Full Product Details