Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tap42
- Entrez Gene ID
-
326166
- Gene Name
- Two A-associated protein of 42kDa
- Gene Aliases
-
CG31852, CG6537, Dmel\CG31852, Dmel_CG31852, Tap-42/CG31852, dTap42
- Chromosome Location
-
Chr.2L: 13367617 - 13369021 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051852
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165039.3 |
NP_723811.1 |
1 - 2 |
227 |
60 |
GenBank mRNA |
BT030986.1 |
- |
1 - 2 |
212 |
60 |
View Full Product Details