Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4
Entrez Gene ID
34767
Gene Name
B4-PA
Gene Aliases
B4/Susi, BG:DS07660.4, CG9239, CG9260, Dmel\CG9239, Dmel_CG9239, Susi, n(2)05337
Chromosome Location
Chr.2L: 13500624 - 13549328 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023407
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273516.1 NP_001260445.1 5 - 6 1556 65
NM_057977.4 NP_477325.2 5 - 6 1414 65
NM_165046.1 NP_723823.1 5 - 6 1237 65
GenBank mRNA AY060445.1 - 5 - 6 1397 65
AY061420.1 - 2 - 3 728 65

View Full Product Details