Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpS23
- Entrez Gene ID
-
318977
- Gene Name
- mitochondrial ribosomal protein S23
- Gene Aliases
-
CG31842, Dmel\CG31842, Dmel_CG31842, l(2)SH0272, l(2)SH2 0272
- Chromosome Location
-
Chr.2L: 13834250 - 13835072 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260407
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165065.4 |
NP_723847.1 |
1 - 2 |
128 |
69 |
View Full Product Details