Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CenG1A
Entrez Gene ID
34803
Gene Name
Centaurin gamma 1A
Gene Aliases
AGAP, BG:DS08220.1, CG16866, CG16867, CG31811, CG7982, CenG1a, Dmel\CG31811, Dmel_CG31811, cenG, cenG1A, cenG1a, ceng1A
Chromosome Location
Chr.2L: 13835564 - 13898712 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078838.3 NP_523562.2 2 - 3 980 94
NM_165067.2 NP_723849.1 2 - 3 1099 94
GenBank mRNA AY122158.1 - 2 - 3 962 94
BT133428.1 - 2 - 3 1096 94

View Full Product Details