Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CenG1A
Entrez Gene ID
34803
Gene Name
Centaurin gamma 1A
Gene Aliases
AGAP, BG:DS08220.1, CG16866, CG16867, CG31811, CG7982, CenG1a, Dmel\CG31811, Dmel_CG31811, cenG, cenG1A, cenG1a, ceng1A
Chromosome Location
Chr.2L: 13835564 - 13898712 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078838.3 NP_523562.2 3 - 4 1121 114
NM_165067.2 NP_723849.1 3 - 4 1240 114
NM_165068.2 NP_723850.1 2 - 3 312 114
GenBank mRNA AY122158.1 - 3 - 4 1103 114
BT053695.1 - 2 - 3 290 114
BT133428.1 - 3 - 4 1237 114

View Full Product Details