Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osp
Entrez Gene ID
34850
Gene Name
outspread
Gene Aliases
BG:DS01486.5, BG:DS01486.7, BG:DS07721.2, CG3479, Dmel\CG3479, Dmel_CG3479, alw, anon-WO02059370.75, l(2)rJ571, v(2)rJ571
Chromosome Location
Chr.2L: 14599196 - 14689340 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003016
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144354.3 NP_001137826.1 7 - 8 4415 78
NM_001169504.2 NP_001162975.1 7 - 8 4415 78
NM_078843.6 NP_523567.5 6 - 7 4388 78
NM_165089.4 NP_723879.3 6 - 7 4388 78
GenBank mRNA BT031256.1 - 6 - 7 4358 78

View Full Product Details