Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ca-alpha1D
Entrez Gene ID
34950
Gene Name
Ca[2+]-channel protein alpha[[1]] subunit D
Gene Aliases
A1D, BG:DS02795.1, CG4894, Ca-Alpha1D, Ca-_1D, Ca-a1D, Ca-alpha1, Ca-alpha[[1]]D, Ca1D, Ca_alpha1D, Caalpha1D, D-type Ca[2+], DmCa1D, DmCa1alpha1, Dmca1D, Dmel\CG4894, Dmel_CG4894, DroCa1, alpha1, alpha1D, ca-alpha1D, ca-alpha1d, caID, cad, l(2)35Fa, l35Fa
Chromosome Location
Chr.2L: 16165831 - 16190540 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001991
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273551.1 NP_001260480.2 14 - 15 4796 83
NM_001273552.1 NP_001260481.1 13 - 14 3703 83
NM_001273553.1 NP_001260482.1 13 - 14 3703 83
NM_001273554.1 NP_001260483.1 13 - 14 3703 83
NM_001273555.1 NP_001260484.1 13 - 14 3703 83
NM_001273556.1 NP_001260485.1 13 - 14 3703 83
NM_080365.3 NP_525104.3 14 - 15 4786 83
NM_134429.3 NP_602305.2 14 - 15 4786 83
NM_165146.2 NP_723952.2 14 - 15 4700 83
NM_165147.2 NP_723953.2 14 - 15 4700 83

View Full Product Details