Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- shakB
- Entrez Gene ID
-
33062
- Gene Name
- shaking B
- Gene Aliases
-
2, 3, 6, CG12678, CG1321, CG15451, CG32508, CG34358, Dmel\CG34358, Dmel_CG34358, E81, FBpp0111481, Pas, R-9-29, R9-29, SHAK-B, Shak B, Shak-B, ShakB, W3, inx8, l(1)19Eb, l(1)LB21, l(1)R-10-14, l(1)R-10-3, l(1)R-10-7, l(1)R-9-29, l(1)TH73, l(1)W3, l(1)W3b, lincRNA.1031, nj-156, nj156, pas, pass, shB, shak-B, shkB
- Chromosome Location
-
Chr.X: 20761071 - 20927050 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0085387
View Full Product Details