Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NfI
Entrez Gene ID
43782
Gene Name
Nuclear factor I
Gene Aliases
CG2380, Dmel\CG2380, Dmel_CG2380, NFI
Chromosome Location
Chr.4: 268175 - 276369 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0042696
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272122.2 NP_001259051.1 7 - 8 2181 126
NM_205870.3 NP_995592.2 7 - 8 2193 126
GenBank mRNA BT003547.1 - 7 - 8 2191 126
BT023941.1 - 7 - 8 2191 126

View Full Product Details