Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtnl1
Entrez Gene ID
33721
Gene Name
Reticulon-like1
Gene Aliases
142893_at, CG18623, CG33113, CG8895, Dmel\CG33113, Dmel_CG33113, G9, Retl1, Rtn1, Rtnl-1, rtnl1
Chromosome Location
Chr.2L: 4992808 - 5009720 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053113
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175972.3 NP_787986.1 2 - 3 281 61
NM_205908.2 NP_995630.1 1 - 2 276 61
GenBank mRNA BT132825.1 - 2 - 3 251 61

View Full Product Details