Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rtnl1
Entrez Gene ID
33721
Gene Name
Reticulon-like1
Gene Aliases
142893_at, CG18623, CG33113, CG8895, Dmel\CG33113, Dmel_CG33113, G9, Retl1, Rtn1, Rtnl-1, rtnl1
Chromosome Location
Chr.2L: 4992808 - 5009720 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053113
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169404.2 NP_001162875.1 5 - 6 2097 68
NM_001169405.2 NP_001162876.1 6 - 7 856 68
NM_001169406.2 NP_001162877.1 6 - 7 839 68
NM_001169407.2 NP_001162878.2 4 - 5 836 68
NM_175969.3 NP_787983.1 4 - 5 2061 68
NM_175970.3 NP_787984.1 6 - 7 900 68
NM_175971.3 NP_787985.1 5 - 6 872 68
NM_175972.3 NP_787986.1 5 - 6 804 68
NM_175973.3 NP_787987.1 4 - 5 789 68
NM_175974.3 NP_787988.1 4 - 5 628 68
NM_205908.2 NP_995630.1 4 - 5 799 68
GenBank mRNA AY069432.1 - 6 - 7 866 68
BT099515.1 - 4 - 5 616 68
BT099529.1 - 4 - 5 785 68
BT132825.1 - 5 - 6 774 68

View Full Product Details