Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nompC
Entrez Gene ID
33768
Gene Name
no mechanoreceptor potential C
Gene Aliases
CG11020, CG17468, CT30855, DmNompC, Dmel\CG11020, Dmel_CG11020, NOMPC, NompC, ORE-13, TRPN, TRPN1, jf24, l(2)25Dc, l(2)jf24, nomp
Chromosome Location
Chr.2L: 5346200 - 5365760 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016920
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103619.4 NP_001097089.2 5 - 6 1440 61
NM_001258962.3 NP_001245891.1 5 - 6 1440 61
NM_001316380.1 NP_001303309.1 5 - 6 1440 61
NM_078759.4 NP_523483.2 5 - 6 1440 61
NM_205912.4 NP_995634.2 5 - 6 1440 61
GenBank mRNA HM582118.1 - 5 - 6 782 61

View Full Product Details