Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AlaRS
Entrez Gene ID
34156
Gene Name
Alanyl-tRNA synthetase
Gene Aliases
ARS, Aats-ala, CG13391, Dmel\CG13391, Dmel_CG13391, alaS, dAARS
Chromosome Location
Chr.2L: 8378262 - 8381764 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027094
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078787.4 NP_523511.2 2 - 3 1085 60
NM_205934.3 NP_995656.1 1 - 2 1136 60
GenBank mRNA BT150285.1 - 1 - 2 1217 60

View Full Product Details