Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcm2
Entrez Gene ID
34280
Gene Name
CG3858-PA
Gene Aliases
CG3858, Dmel\CG3858, Dmel_CG3858, FBgn0019809, GCM-2, Gcm2, dGCM2
Chromosome Location
Chr.2L: 9608562 - 9612706 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0019809
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135458.5 NP_609302.1 3 - 4 1183 73
GenBank mRNA AF184664.1 - 3 - 4 1207 73
BT022152.1 - 3 - 4 1096 73

View Full Product Details