The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038817.1 | NP_001033906.1 | 9 - 10 | 1570 | 82 |
NM_001038818.1 | NP_001033907.1 | 9 - 10 | 1743 | 82 | |
NM_001273526.1 | NP_001260455.1 | 9 - 10 | 1816 | 82 | |
NM_001273527.1 | NP_001260456.1 | 5 - 6 | 1163 | 82 | |
NM_078840.5 | NP_523564.1 | 8 - 9 | 1738 | 82 | |
NM_205988.3 | NP_995710.1 | 9 - 10 | 1834 | 82 | |
NM_205989.3 | NP_995711.1 | 9 - 10 | 1830 | 82 | |
GenBank mRNA | AF168467.1 | - | 9 - 10 | 1805 | 82 |
BT010088.1 | - | 8 - 9 | 1738 | 82 | |
BT133412.1 | - | 5 - 6 | 1109 | 82 |