Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fbw5
- Entrez Gene ID
-
33854
- Gene Name
- F-box and WD repeat domain containing 5
- Gene Aliases
-
CG9144, Dmel\CG9144, Dmel_CG9144, FBXW2, FBgn 31773, dFbw5
- Chromosome Location
-
Chr.2L: 6063562 - 6065953 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031773
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135145.2 |
NP_608989.1 |
2 - 3 |
2078 |
65 |
GenBank mRNA |
AY118759.1 |
- |
2 - 3 |
2046 |
65 |
View Full Product Details