Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TTLL3A
- Entrez Gene ID
-
33947
- Gene Name
- Tubulin tyrosine ligase-like 3A
- Gene Aliases
-
CG11323, DmCG11323, Dmel\CG11323, Dmel_CG11323, TTLL3, dmTTLL3A
- Chromosome Location
-
Chr.2L: 6736364 - 6740188 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031854
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135225.2 |
NP_609069.1 |
3 - 4 |
1901 |
56 |
View Full Product Details