Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Wnt10
- Entrez Gene ID
-
34011
- Gene Name
- Wnt oncogene analog 10
- Gene Aliases
-
CG4971, D-Wnt-10, DWnt-10, DWnt10, Dm DWnt10, Dmel\CG4971, Dmel_CG4971, Dwnt-10, Dwnt10, wnt10
- Chromosome Location
-
Chr.2L: 7363379 - 7377528 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031903
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135265.5 |
NP_609109.3 |
5 - 6 |
1457 |
71 |
View Full Product Details