Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ntl
- Entrez Gene ID
-
34046
- Gene Name
- Neurotransmitter transporter-like
- Gene Aliases
-
CG7075, Dmel\CG7075, Dmel_CG7075, Mp, ntl
- Chromosome Location
-
Chr.2L: 7710880 - 7713991 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0267326
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135291.3 |
NP_609135.1 |
1 - 2 |
327 |
68 |
View Full Product Details