Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr82
Entrez Gene ID
34153
Gene Name
WD repeat domain 82
Gene Aliases
CG17293, Dmel\CG17293, Dmel_CG17293, WDR82, dWdr82, wdr-82, wdr82
Chromosome Location
Chr.2L: 8374461 - 8375549 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032030
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135373.3 NP_609217.1 1 - 1 944 100
GenBank mRNA AY069081.1 - 1 - 1 924 100

View Full Product Details