Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ppk18
- Entrez Gene ID
-
34295
- Gene Name
- pickpocket 18
- Gene Aliases
-
CG13120, CG13121, CG44152, Dmel\CG44152, Dmel_CG44152
- Chromosome Location
-
Chr.2L: 9765357 - 9770996 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265001
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135464.3 |
NP_609308.3 |
4 - 5 |
832 |
96 |
View Full Product Details