Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IP3K1
Entrez Gene ID
34300
Gene Name
Inositol 1,4,5-triphosphate kinase 1
Gene Aliases
CG4026, D-IP3K1, Dmel\CG4026, Dmel_CG4026, IP3KI, IP[[3]]K-1, Ip3k1, UY530, anon-WO0118547.189, dmIP3Kalpha, dmIP[[3]]Kalpha
Chromosome Location
Chr.2L: 9782384 - 9788499 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032147
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259025.1 NP_001245954.1 3 - 4 271 69
NM_135469.4 NP_609313.2 3 - 4 909 69

View Full Product Details