Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- obst-B
- Entrez Gene ID
-
34336
- Gene Name
- obstructor-B
- Gene Aliases
-
BEST:CK00178, BcDNA:GH02976, CG4778, CK00178, Dmel\CG4778, Dmel_CG4778
- Chromosome Location
-
Chr.2L: 10032690 - 10038954 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027600
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135495.3 |
NP_609339.1 |
2 - 3 |
115 |
71 |
GenBank mRNA |
AF145612.1 |
- |
2 - 3 |
98 |
71 |
View Full Product Details