Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klp31E
Entrez Gene ID
34422
Gene Name
Kinesin-like protein at 31E
Gene Aliases
CG5300, DmCG5300, DmKlp31D, DmKlp31E, Dmel\CG5300, Dmel_CG5300, KIF 21A, KIF21A/B, KLP31E
Chromosome Location
Chr.2L: 10414017 - 10418991 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032243
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169464.1 NP_001162935.1 8 - 9 2068 80
NM_001169465.2 NP_001162936.1 9 - 10 2169 80
NM_001169466.2 NP_001162937.2 9 - 10 2186 80
NM_135554.3 NP_609398.2 9 - 10 2189 80
GenBank mRNA BT001471.1 - 9 - 10 2172 80

View Full Product Details