Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RluA-2
Entrez Gene ID
34440
Gene Name
RluA pseudouridine synthase 2
Gene Aliases
CG6187, Dmel\CG6187, Dmel_CG6187, yumo
Chromosome Location
Chr.2L: 10477073 - 10482975 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032256
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201853.2 NP_001188782.2 2 - 3 381 105
NM_001201854.2 NP_001188783.1 1 - 2 514 105
NM_001273423.1 NP_001260352.1 1 - 2 514 105
NM_135567.4 NP_609411.1 1 - 2 514 105
GenBank mRNA AY058686.1 - 1 - 2 466 105

View Full Product Details