Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csl4
Entrez Gene ID
34548
Gene Name
CG6249-PA
Gene Aliases
CG6249, CSL4, Dmel\CG6249, Dmel_CG6249, dCsl4
Chromosome Location
Chr.2L: 11122574 - 11123567 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032346
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135648.2 NP_609492.1 1 - 2 256 70
GenBank mRNA AY095082.1 - 2 - 3 422 70

View Full Product Details