Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr36b
- Entrez Gene ID
-
117487
- Gene Name
- Gustatory receptor 36b
- Gene Aliases
-
CG15137, CG31744, Dmel\CG31744, Dmel_CG31744, GR36b, Gr36B.2, Gr36B2
- Chromosome Location
-
Chr.2L: 17178772 - 17180064 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045486
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165206.1 |
NP_724039.1 |
2 - 3 |
1026 |
77 |
View Full Product Details