Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dif
Entrez Gene ID
35045
Gene Name
Dorsal-related immunity factor
Gene Aliases
6794, CG6794, DIF, Dif2, Dmel\CG6794, Dmel_CG6794, dif
Chromosome Location
Chr.2L: 17413248 - 17433161 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011274
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169528.1 NP_001162999.1 6 - 7 1506 74
NM_078865.4 NP_523589.2 6 - 7 1746 74
NM_165216.3 NP_724050.1 6 - 7 1394 74
GenBank mRNA BT025231.1 - 6 - 7 1390 74

View Full Product Details